Ethylenediamine-N,N'-dipropionic acid dihydrochloride
Supplier: TCI America
E0238-1G
E0238-25G
TCE0238-025GEA
239.43
USD
TCE0238-025G
TCE0238-001G
Ethylenediamine-N,N'-dipropionic acid dihydrochloride
Ethylenediamine-N,N'-dipropionic acid dihydrochloride
CAS Number: 32705-91-2
MDL Number: MFCD00060168
Molecular Formula: C8H16N2O4
Molecular Weight: 277.14
Form: Crystal
Color: White
Melting point (°C): 198
Formula:
C₈H₁₆N₂O₄ Melting Pt: 198 °C |
MDL Number:
MFCD00060168 CAS Number: 32705-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...