Eriochrome black A
Synonyms:
Chrome Black A
E0226-25G
TCE0226-025GEA
60.77
USD
TCE0226-025G
Eriochrome black A
Eriochrome black A
CAS Number: 3618-58-4
MDL Number: MFCD00054390
Molecular Formula: C20H13N3O7S
Molecular Weight: 461.38
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: Chrome Black A
Formula:
C₂₀H₁₃N₃O₇S MW: 461.38 g/mol |
MDL Number:
MFCD00054390 CAS Number: 3618-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...