α-Chloro-4-ethyltoulene 65.0% (by GC) (contains ca. 30% 2-form)

Danger

Synonyms: 1-(Chloromethyl)-4-ethylbenzeneα-Chloro-p-ethyltoulene4-Ethylbenzyl chloride

E0215-25G
TCE0215-025GEA 54.39 USD
TCE0215-025G
α-Chloro-4-ethyltoulene 65.0% (by GC) (contains ca. 30% 2-form)
α-Chloro-4-ethyltoulene

CAS Number: 1467-05-6
MDL Number: MFCD00039357
Molecular Formula: C9H11Cl
Molecular Weight: 154.64
Purity/Analysis Method: 65.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 1.05

Formula: C₉H₁₁Cl
MW: 154.64 g/mol
Boiling Pt: 111 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00039357
CAS Number: 1467-05-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Purity/Analysis Method 65.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR