Eriochrome cyanine R

Supplier: TCI America

Synonyms: Chromoxane Cyanine R

E0201-25G
TCE0201-025GEA 54.96 USD InStock
TCE0201-025G
Eriochrome cyanine R
Eriochrome cyanine R

CAS Number: 3564-18-9
MDL Number: MFCD00012411
Molecular Formula: C23H18O9S
Molecular Weight: 536.39
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Solochrome Cyanine R, Mordant Blue 3

Formula: C₂₃H₁₅Na₃O₉S
MDL Number: MFCD00012411
CAS Number: 3564-18-9
EINECS: 222-641-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR