TCIAA1272

A1272-1G A1272-5G
TCA1272-5GEA 505.88 USD
TCA1272-5G TCA1272-001G
TCIAA1272
CAS Number: 888-54-0
MDL Number: MFCD00191599
Molecular Formula: C9H8N4O4
Molecular Weight: 236.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 165

Other Product Names / Synonyms: 2-Propenal 2,4-Dinitrophenylhydrazone
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR