N-Ethylrhodanine ≥98.0%

Supplier: TCI America

Synonyms: 3-Ethylrhodanine3-Ethyl-2-thioxothiazolidin-4-one

E0174-25G E0174-5G
TCE0174-5GEA 74.1 USD
TCE0174-5G TCE0174-025G
N-Ethylrhodanine ≥98.0%
N-Ethylrhodanine

CAS Number: 7648-01-3
MDL Number: MFCD00051339
Molecular Formula: C5H7NOS2
Molecular Weight: 161.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 128
Melting point (°C): 37
Storage Temperature: 0-10°C

Formula: C₅H₇NOS₂
MW: 161.25 g/mol
Boiling Pt: 128 °C
Melting Pt: 37 °C
MDL Number: MFCD00051339
CAS Number: 7648-01-3
EINECS: 231-603-9
https://us.vwr.com/stibo/web/std.lang.all/77/49/28967749.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR