TCIAE0123

E0123-25ML E0123-500ML
TCE0123-025MLEA 39.36 USD
TCE0123-025ML TCE0123-500ML
TCIAE0123
CAS Number: 645-62-5
MDL Number: MFCD00006983
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 68
Specific Gravity (20/20): 0.85
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR