Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate

Supplier: TCI America

Synonyms: Nickel(II) Disodium Ethylenediaminetetraacetate

E0097-25G
TCE0097-025GEA 304.7 USD
TCE0097-025G
Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate
Ethylenediaminetetraacetic acid disodium nickel(II) salt hydrate

CAS Number: 15708-55-1
MDL Number: MFCD00070493
Molecular Formula: C10H16N2O8
Molecular Weight: 392.88
Form: Crystal
Color: Pale Blue


Other Product Names / Synonyms: Nickel(II) Disodium Ethylenediaminetetraacetate

Formula: C₁₀H₁₆N₂O₈
MW: 392.88 g/mol
MDL Number: MFCD00070493
CAS Number: 15708-55-1
https://us.vwr.com/stibo/web/std.lang.all/76/93/28967693.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR