Ethylenediaminetetraacetic acid disodium cobalt salt hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Cobalt Disodium Ethylenediaminetetraacetate
E0089-25G
TCE0089-025GEA
112.64
USD
TCE0089-025G
Ethylenediaminetetraacetic acid disodium cobalt salt hydrate ≥98.0% (by titrimetric analysis)
Ethylenediaminetetraacetic acid disodium cobalt salt hydrate
CAS Number: 15137-09-4
MDL Number: MFCD00054448
Molecular Formula: C10H16N2O8
Molecular Weight: 393.12
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Cobalt Disodium Ethylenediaminetetraacetate
Formula:
C₁₀H₁₆N₂O₈ MW: 393.12 g/mol |
MDL Number:
MFCD00054448 CAS Number: 15137-09-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...