Ethyl propyl ether ≥98.0%
Supplier: TCI America
Synonyms:
3-Ethoxypropane
E0052-25ML
TCE0052-025MLEA
188.21
USD
TCE0052-025ML
Ethyl propyl ether ≥98.0%
Ethyl propyl ether
CAS Number: 628-32-0
MDL Number: MFCD00015215
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): -20
Specific Gravity (20/20): 0.74
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Ethoxypropane
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 62 °C Density: 0.74 (20 °C) Flash Pt: –20 °C |
MDL Number:
MFCD00015215 CAS Number: 628-32-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...