(±)-Epoxystyrene ≥98.0%

Supplier: TCI America

Synonyms: Styrene oxide

E0013-500ML E0013-25ML
TCE0013-500MLEA 62.27 USD
TCE0013-500ML TCE0013-025ML
(±)-Epoxystyrene ≥98.0%
(±)-Epoxystyrene

CAS Number: 96-09-3
MDL Number: MFCD00005121
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Melting point (°C): -37
Flash Point (°C): 77
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Epoxyethylbenzene, Phenylethylene Oxide, Phenyloxirane

Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 194 °C
Melting Pt: –37 °C
Density: 1.06 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00005121
CAS Number: 96-09-3
EINECS: 202-476-7
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/76/26/28967626.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR