Dimesitylfluoroborane ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Dimesitylboron Fluoride
D3890-1G
D3890-5G
TCD3890-5GEA
177.31
USD
TCD3890-5G
TCD3890-1G
Dimesitylfluoroborane ≥98.0% (by HPLC)
Dimesitylfluoroborane
CAS Number: 436-59-9
MDL Number: MFCD00012203
Molecular Formula: C18H22BF
Molecular Weight: 268.18
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 129
Melting point (°C): 74
Other Product Names / Synonyms: Dimesitylboron Fluoride
Formula:
C₁₈H₂₂BF MW: 268.18 g/mol Boiling Pt: 129 °C Melting Pt: 74 °C |
MDL Number:
MFCD00012203 CAS Number: 436-59-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...