4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Pyrimidine-4,5-diamine

D3858-1G
TCD3858-1GEA 183.72 USD
TCD3858-1G
4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
4,5-Diaminopyrimidine

CAS Number: 13754-19-3
MDL Number: MFCD00006113
Molecular Formula: C4H6N4
Molecular Weight: 110.12
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 229
Melting point (°C): 203

https://us.vwr.com/stibo/web/std.lang.all/58/08/28985808.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR