Diammonium dithiodiglycolate 45% in water

Synonyms: Diammonium DithiodiacetateDithiodiglycolic Acid Diammonium SaltDithiodiacetic Acid Diammonium Salt

D3829-500G
TCD3829-500GEA 158.1 USD
TCD3829-500G
Diammonium dithiodiglycolate 45% in water
Diammonium dithiodiglycolate

CAS Number: 68223-93-8
MDL Number: MFCD00072606
Molecular Formula: C4H12N2O4S2
Molecular Weight: 216.27
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: Dithiodiacetic Acid Diammonium Salt, Diammonium Dithiodiacetate, Dithiodiglycolic Acid Diammonium Salt

Formula: C₄H₁₂N₂O₄S₂
MW: 216.27 g/mol
Density: 1.21 (20 °C)
MDL Number: MFCD00072606
CAS Number: 68223-93-8
https://us.vwr.com/stibo/web/std.lang.all/57/89/28985789.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR