9,9-Dimethylfluorene ≥99.0%, purified by sublimation

Supplier: TCI America

Synonyms: 9,9-Dimethyl-9H-fluorene

D3810-1G
TCD3810-1GEA 113.68 USD
TCD3810-1G
9,9-Dimethylfluorene ≥99.0%, purified by sublimation
9,9-Dimethylfluorene

CAS Number: 4569-45-3
MDL Number: MFCD00114670
Molecular Formula: C15H14
Molecular Weight: 194.28
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 287
Melting point (°C): 97
Lambda max.: 301 nm (EtOH)

Formula: C₁₅H₁₄
MW: 194.28 g/mol
Boiling Pt: 287 °C
Melting Pt: 97 °C
MDL Number: MFCD00114670
CAS Number: 4569-45-3
https://us.vwr.com/stibo/web/std.lang.all/57/73/28985773.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR