(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0%
Supplier: TCI America
Synonyms:
(1S,2S)-N1,N2-Bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine
D3809-1G
TCD3809-1GEA
201.92
USD
TCD3809-1G
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0%
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine
CAS Number: 767291-67-8
Molecular Formula: C20H42N2
Molecular Weight: 310.57
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 0.87
Specific rotation [a]20/D: 34 deg (C=1, CHCl3)
Formula:
C₂₀H₄₂N₂ MW: 310.57 g/mol Boiling Pt: 145 °C Density: 0,87 (20 °C) |
MDL Number:
MFCD11869717 CAS Number: 767291-67-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...