(S)-(-)-2-(Diphenylmethyl)pyrrolidine ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-2-Diphenylmethylpyrrolidine

D3804-1G
TCD3804-1GEA 241.49 USD
TCD3804-1G
(S)-(-)-2-(Diphenylmethyl)pyrrolidine ≥95.0% (by GC, titration analysis)
(S)-(-)-2-(Diphenylmethyl)pyrrolidine

CAS Number: 119237-64-8
MDL Number: MFCD00799525
Molecular Formula: C17H19N
Molecular Weight: 237.35
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 113
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: -3 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₁₇H₁₉N
MW: 237.35 g/mol
Boiling Pt: 135 °C
Density: 1.06 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00799525
CAS Number: 119237-64-8
https://us.vwr.com/stibo/web/std.lang.all/57/68/28985768.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR