3-(Dimethylsilyloxy)-1,1,5,5-tetramethyl-3-phenyltrisiloxane ≥96.0%

Supplier: TCI America

Synonyms: Tris(dimethylsilyloxy)phenylsilanePhenyltris(dimethylsiloxy)silane

D3762-25ML
TCD3762-25MLEA 163 USD
TCD3762-25ML
3-(Dimethylsilyloxy)-1,1,5,5-tetramethyl-3-phenyltrisiloxane ≥96.0%
3-(Dimethylsilyloxy)-1,1,5,5-tetramethyl-3-phenyltrisiloxane

CAS Number: 18027-45-7
MDL Number: MFCD00053849
Molecular Formula: C12H26O3Si4
Molecular Weight: 330.68
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 87
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Tris(dimethylsilyloxy)phenylsilane

Formula: C₁₂H₂₆O₃Si₄
MW: 330.68 g/mol
Boiling Pt: 91 °C
Density: 0.94 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00053849
CAS Number: 18027-45-7
https://us.vwr.com/stibo/web/std.lang.all/57/31/28985731.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR