Di-1-naphthylmethanol ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Di(naphthalen-1-yl)methanolBis(alpha-naphthyl)methanol

D3750-5G
TCD3750-5GEA 162.35 USD
TCD3750-5G
Di-1-naphthylmethanol ≥98.0% (by HPLC)
Di-1-naphthylmethanol

CAS Number: 62784-66-1
Molecular Formula: C21H16O
Molecular Weight: 284.36
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: Bis(alpha-naphthyl)methanol

Formula: C₂₁H₁₆O
MW: 284.36 g/mol
Melting Pt: 148 °C
MDL Number: MFCD11096937
CAS Number: 62784-66-1
https://us.vwr.com/stibo/web/std.lang.all/57/17/28985717.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR