Diclofenac ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Diclofenac acid

D3748-25G
TCD3748-25GEA 64.72 USD
TCD3748-25G
Diclofenac ≥98.0% (by GC, titration analysis)
Diclofenac

CAS Number: 15307-86-5
Molecular Formula: C14H11Cl2NO2
Molecular Weight: 296.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 158


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: Diclofenac

Formula: C₁₄H₁₁Cl₂NO₂
Melting Pt: 158 °C
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/57/15/28985715.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR