Dibenzo[g,p]chrysene ≥98.0% (by HPLC)

Supplier: TCI America

D3736-200MG D3736-1G
TCD3736-200MGEA 146.09 USD
TCD3736-200MG TCD3736-1G
Dibenzo[g,p]chrysene ≥98.0% (by HPLC)
Dibenzo[g,p]chrysene

CAS Number: 191-68-4
Molecular Formula: C26H16
Molecular Weight: 328.41
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 218

Formula: C₂₆H₁₆
Melting Pt: 218 °C
CAS Number: 191-68-4
https://us.vwr.com/stibo/web/std.lang.all/57/07/28985707.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR