1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane ≥90.0%

D3734-5G
TCD3734-5GEA 199.82 USD
TCD3734-5G
1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane ≥90.0%
1,3-Dimethoxy-1,1,3,3-tetraphenyldisiloxane

CAS Number: 94593-08-5
MDL Number: MFCD05663939
Molecular Formula: C26H26O3Si2
Molecular Weight: 442.66
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Boiling point (°C): 210

Formula: C₂₆H₂₆O₃Si₂
Boiling Pt: 210 °C
MDL Number: MFCD05663939
CAS Number: 94593-08-5
https://us.vwr.com/stibo/web/std.lang.all/57/06/28985706.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR