Dimethoxydi-p-tolylsilane ≥98.0%

Supplier: TCI America

D3733-1G D3733-5G
TCD3733-5GEA 301 USD
TCD3733-5G TCD3733-1G
Dimethoxydi-p-tolylsilane ≥98.0%
Dimethoxydi-p-tolylsilane

CAS Number: 92779-72-1
MDL Number: MFCD01861714
Molecular Formula: C16H20O2Si
Molecular Weight: 272.42
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.05

Formula: C₁₆H₂₀O₂Si
Density: 1.05 (20 °C)
MDL Number: MFCD01861714
CAS Number: 92779-72-1
https://us.vwr.com/stibo/web/std.lang.all/57/05/28985705.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR