Diethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
cis-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester
D3714-5G
TCD3714-5GEA
194.79
USD
TCD3714-5G
Diethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥98.0%
Diethyl-cis-4-cyclohexene-1,2-dicarboxylate
CAS Number: 4841-85-4
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 114
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester
Formula:
C₁₂H₁₈O₄ MW: 226.27 g/mol Boiling Pt: 114 °C Density: 1.08 (20 °C) |
CAS Number:
4841-85-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...