Diethyl-cis-1,2-cyclohexanedicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: cis-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester

D3713-5G
TCD3713-5GEA 399 USD
TCD3713-5G
Diethyl-cis-1,2-cyclohexanedicarboxylate ≥98.0%
Diethyl-cis-1,2-cyclohexanedicarboxylate

CAS Number: 17351-07-4
MDL Number: MFCD03938293
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: cis-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Boiling Pt: 119 °C
Density: 1.06 (20 °C)
MDL Number: MFCD03938293
CAS Number: 17351-07-4
https://us.vwr.com/stibo/web/std.lang.all/56/91/28985691.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR