1,5-Dioxepan-2-one ≥98.0%

Supplier: TCI America

D3707-25G D3707-5G
TCD3707-5GEA 308 USD
TCD3707-5G TCD3707-25G
1,5-Dioxepan-2-one ≥98.0%
1,5-Dioxepan-2-one

CAS Number: 35438-57-4
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 36
Storage Temperature: 0°C

https://us.vwr.com/stibo/web/std.lang.all/56/84/28985684.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR