Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D3706-100MG
TCD3706-100MGEA
279.05
USD
TCD3706-100MG
Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate ≥97.0% (by titrimetric analysis)
Dihydrogen di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate
CAS Number: 386706-31-6
MDL Number: MFCD05662696
Molecular Formula: C32H74Cl2O4P4Pd2
Molecular Weight: 930.58
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 280
Formula:
C₃₂H₇₄Cl₂O₄P₄Pd₂ Melting Pt: 280 °C |
MDL Number:
MFCD05662696 CAS Number: 386706-31-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...