Acid red 13

Supplier: TCI America

Synonyms: Fast Red E

A1243-10G
TCA1243-010GEA 80.15 USD
TCA1243-010G
Acid red 13
Acid red 13

CAS Number: 2302-96-7
MDL Number: MFCD00191366
Molecular Formula: C20H14N2O7S2
Molecular Weight: 502.42
Form: Crystal
Color: Deep Red
Flash Point (°C): 93
Lambda max.: 505 nm (0.02mol/L AcONH4 sol.)


Other Product Names / Synonyms: Fast Red E

Formula: C₂₀H₁₄N₂O₇S₂
MW: 502.42 g/mol
Flash Pt: 93 °C
MDL Number: MFCD00191366
CAS Number: 2302-96-7
https://us.vwr.com/stibo/web/std.lang.all/91/06/28929106.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR