Diethyl-trans-1,2-cyclohexanedicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
trans-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester
D3684-5G
TCD3684-5GEA
488
USD
TCD3684-5G
Diethyl-trans-1,2-cyclohexanedicarboxylate ≥98.0%
Diethyl-trans-1,2-cyclohexanedicarboxylate
CAS Number: 17351-22-3
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 104
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: trans-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester
Formula:
C₁₂H₂₀O₄ MW: 228.29 g/mol Boiling Pt: 104 °C Density: 1.04 (20 °C) |
CAS Number:
17351-22-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...