1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-[2-(2,4-Dichlorophenyl)-2-hydroxyethyl]imidazole
, 1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol
D3629-25G
TCD3629-25GEA
82.13
USD
TCD3629-25G
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol ≥98.0% (by GC, titration analysis)
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
CAS Number: 24155-42-8
MDL Number: MFCD00044708
Molecular Formula: C11H10Cl2N2O
Molecular Weight: 257.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 135
Other Product Names / Synonyms: 1-[2-(2,4-Dichlorophenyl)-2-hydroxyethyl]imidazole
Formula:
C₁₁H₁₀Cl₂N₂O MW: 257.11 g/mol Melting Pt: 135 °C |
MDL Number:
MFCD00044708 CAS Number: 24155-42-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...