1,3-Diisopropylimidazolinium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: IiPrim.HBF4

D3622-1G D3622-5G
TCD3622-1GEA 272.19 USD
TCD3622-1G TCD3622-5G
1,3-Diisopropylimidazolinium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1,3-Diisopropylimidazolinium tetrafluoroborate

CAS Number: 137581-18-1
MDL Number: MFCD08276803
Molecular Formula: C9H19BF4N2
Molecular Weight: 242.07
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 146


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: IiPrim.HBF4

Formula: C₉H₁₉BF₄N₂
MW: 242.07 g/mol
Melting Pt: 146 °C
MDL Number: MFCD08276803
CAS Number: 137581-18-1
https://us.vwr.com/stibo/web/std.lang.all/56/12/28985612.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR