2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine ≥98.0% (by total nitrogen basis) (contains o-form)

Supplier: TCI America

Synonyms: 5-Aminospiro[naphthalene-1(4H),2'(3'H)-[1H]perimidine]-4-onePNO-p

D3618-100MG
TCD3618-100MGEA 1157 USD
TCD3618-100MG
2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine ≥98.0% (by total nitrogen basis) (contains o-form)
2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine

CAS Number: 851768-63-3
Molecular Formula: C20H15N3O
Molecular Weight: 313.36
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 5-Aminospiro[naphthalene-1(4H),2'(3'H)-[1H]perimidine]-4-one, PNO-p

Formula: C₂₀H₁₅N₃O
MW: 313.36 g/mol
CAS Number: 851768-63-3
https://us.vwr.com/stibo/web/std.lang.all/56/07/28985607.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR