Adrenalone hydrochloride, hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3′,4′-Dihydroxy-2-(methylamino)acetophenone hydrochloride
A1234-25G
A1234-5G
TCA1234-5GEA
44.32
USD
TCA1234-5G
TCA1234-025G
Adrenalone hydrochloride, hydrate ≥98.0% (by HPLC, titration analysis)
Adrenalone hydrochloride
Formula:
C₉H₁₂ClNO₃·1H₂O MW: 217.65 g/mol (anhy) |
MDL Number:
MFCD00150331 CAS Number: 62-13-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...