TCIAA1233

A1233-1G
TCA1233-001GEA 80.37 USD
TCA1233-001G
TCIAA1233
CAS Number: 55726-22-2
MDL Number: MFCD00056235
Molecular Formula: C40H78O3
Molecular Weight: 607.06
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 77

Other Product Names / Synonyms: Eicosanoic Anhydride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR