TCIAD3459

D3459-5G D3459-25G
TCD3459-5GEA 23.94 USD
TCD3459-5G TCD3459-25G
TCIAD3459
CAS Number: 3621-82-7
MDL Number: MFCD07368635
Molecular Formula: C7H3Cl2NO
Molecular Weight: 188.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 50
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR