5-(Difluoromethoxy)-2-mercaptobenzimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-(Difluoromethoxy)-1H-benzo[d]imidazole-2-thiol5-(Difluoromethoxy)-2-benzimidazolethiol5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

D3438-25G
TCD3438-25GEA 36 USD
TCD3438-25G
5-(Difluoromethoxy)-2-mercaptobenzimidazole ≥98.0% (by HPLC, titration analysis)
5-(Difluoromethoxy)-2-mercaptobenzimidazole

CAS Number: 97963-62-7
MDL Number: MFCD00467504
Molecular Formula: C8H6F2N2OS
Molecular Weight: 216.21
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 243


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Difluoromethoxy)-2-benzimidazolethiol

Formula: C₈H₆F₂N₂OS
MW: 216.21 g/mol
Melting Pt: 243 °C
MDL Number: MFCD00467504
CAS Number: 97963-62-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR