cis-3,3,5-Trimethylcyclohexyl acetate ≥97.0%
Supplier: TCI America
Synonyms:
Acetic Acid cis-3,3,5-Trimethylcyclohexyl Ester
A1224-25ML
TCA1224-025MLEA
45
USD
TCA1224-025ML
cis-3,3,5-Trimethylcyclohexyl acetate ≥97.0%
cis-3,3,5-Trimethylcyclohexyl acetate
CAS Number: 24691-16-5
MDL Number: MFCD00036661
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 85
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: Acetic Acid cis-3,3,5-Trimethylcyclohexyl Ester
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol Boiling Pt: 97 °C Density: 0.92 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00036661 CAS Number: 24691-16-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...