2',4'-Dichlorovalerophenone ≥95.0%

Supplier: TCI America

Synonyms: 1-(2,4-Dichlorophenyl)pentan-1-one

D3366-5G D3366-25G
TCD3366-25GEA 57.82 USD
TCD3366-25G TCD3366-5G
2',4'-Dichlorovalerophenone ≥95.0%
2',4'-Dichlorovalerophenone

CAS Number: 61023-66-3
Molecular Formula: C11H12Cl2O
Molecular Weight: 231.12
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: 2',4'-Dichloropentanophenone

Formula: C₁₁H₁₂Cl₂O
MW: 231.1184 g/mol
Density: 1.20 (20 °C)
MDL Number: MFCD09028061
CAS Number: 61023-66-3
https://us.vwr.com/stibo/web/std.lang.all/03/52/28980352.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR