Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (Diphenylphosphoryl)(mesityl)methanone

D3358-25G
TCD3358-25GEA 102.6 USD
TCD3358-25G
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide ≥98.0% (by HPLC)
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

CAS Number: 75980-60-8
MDL Number: MFCD00192110
Molecular Formula: C22H21O2P
Molecular Weight: 348.38
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 93
Lambda max.: 400 nm (DMF)

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00192110
CAS Number: 75980-60-8
https://us.vwr.com/stibo/web/std.lang.all/03/46/28980346.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR