Diethyl (4-isopropylbenzyl)phosphonate ≥98.0%

Supplier: TCI America

D3325-5G
TCD3325-5GEA 331 USD
TCD3325-5G
Diethyl (4-isopropylbenzyl)phosphonate ≥98.0%
Diethyl (4-isopropylbenzyl)phosphonate

CAS Number: 77237-55-9
Molecular Formula: C14H23O3P
Molecular Weight: 270.31
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.05


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 4-(Diethylphosphonomethyl)cumene, 1-(Diethylphosphonomethyl)-4-isopropylbenzene, (4-Isopropylbenzyl)phosphonic Acid Diethyl Ester

https://us.vwr.com/stibo/web/std.lang.all/03/09/28980309.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR