1,3-Dibromo-5-(trifluoromethoxy)benzene ≥98.0%
Supplier: TCI America
D3322-5G
D3322-25G
TCD3322-5GEA
32
USD
TCD3322-5G
TCD3322-25G
1,3-Dibromo-5-(trifluoromethoxy)benzene ≥98.0%
1,3-Dibromo-5-(trifluoromethoxy)benzene
CAS Number: 207226-31-1
Molecular Formula: C7H3Br2F3O
Molecular Weight: 319.90
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 113
Specific Gravity (20/20): 1.96
Formula:
C₇H₃Br₂F₃O MW: 319.90 g/mol Boiling Pt: 85 °C Density: 1.96 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00075280 CAS Number: 207226-31-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...