Dibenzylcyanamide ≥98.0%

Supplier: TCI America

Synonyms: N-CyanodibenzylamineN,N-Dibenzylcyanamide

D3320-5G
TCD3320-5GEA 145 USD
TCD3320-5G
Dibenzylcyanamide ≥98.0%
Dibenzylcyanamide

CAS Number: 2451-91-4
Molecular Formula: C15H14N2
Molecular Weight: 222.29
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 54


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: N-Cyanodibenzylamine

Formula: C₁₅H₁₄N₂
MW: 222.29 g/mol
Boiling Pt: 148 °C
Melting Pt: 54 °C
MDL Number: MFCD00094001
CAS Number: 2451-91-4
https://us.vwr.com/stibo/web/std.lang.all/02/95/28980295.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR