1,2-Dihydroxyindan ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Indandiol
D3311-5G
D3311-1G
76219-024EA
16.24
USD
76219-024
TCD3311-5G
1,2-Dihydroxyindan ≥98.0%
1,2-Dihydroxyindan
CAS Number: 4370-02-9
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 158
Other Product Names / Synonyms: 1,2-Indandiol
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Melting Pt: 158 °C |
CAS Number:
4370-02-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...