cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D3309-25G
TCD3309-25GEA
82.18
USD
TCD3309-25G
cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic acid ≥98.0% (by titrimetric analysis)
cis-1,3-Dibenzyl-2-oxo-4,5-imidazolidinedicarboxylic acid
CAS Number: 51591-75-4
Molecular Formula: C19H18N2O5
Molecular Weight: 354.36
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 175
Formula:
C₁₉H₁₈N₂O₅ Melting Pt: 175 °C |
CAS Number:
51591-75-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...