3-Pyridyl acetate ≥98.0%
Supplier: TCI America
Synonyms:
3-Pyridyl acetate
, 3-Acetoxypyridine
, Pyridin-3-yl acetate
A1210-25G
TCA1210-025GEA
132
USD
TCA1210-025G
3-Pyridyl acetate ≥98.0%
3-Pyridyl acetate
CAS Number: 17747-43-2
MDL Number: MFCD00006377
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 100
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 3-Pyridyl Acetate
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 137 °C Density: 1.15 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00006377 CAS Number: 17747-43-2 EINECS: 241-742-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...