9,9-Di-n-octylfluorene ≥97.0% (by HPLC)

Supplier: TCI America

D3285-5G
TCD3285-5GEA 155 USD
TCD3285-5G
9,9-Di-n-octylfluorene ≥97.0% (by HPLC)
9,9-Di-n-octylfluorene

CAS Number: 123863-99-0
Molecular Formula: C29H42
Molecular Weight: 390.66
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Freezing point (°C): 23

Formula: C₂₉H₄₂
CAS Number: 123863-99-0
https://us.vwr.com/stibo/web/std.lang.all/02/60/28980260.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR