1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,10-Bis[4-aza-1-azoniabicyclo[2.2.2]octan-1-yl]decane Dibromide

D3269-1G
TCD3269-1GEA 129 USD
TCD3269-1G
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide

CAS Number: 94630-53-2
Molecular Formula: C22H44Br2N4
Molecular Weight: 524.43
Purity/Analysis Method: 95.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1,10-Bis[4-aza-1-azoniabicyclo[2.2.2]octan-1-yl]decane Dibromide

Formula: C₂₂H₄₄Br₂N₄
MW: 524.43 g/mol
CAS Number: 94630-53-2
https://us.vwr.com/stibo/web/std.lang.all/02/53/28980253.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR