1,4-Dihydro-6-nitroquinoxaline-2,3-dione ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 6-Nitro-1,4-dihydroquinoxaline-2,3-dione

D3241-5G
TCD3241-5GEA 177 USD
TCD3241-5G
1,4-Dihydro-6-nitroquinoxaline-2,3-dione ≥95.0% (by HPLC)
1,4-Dihydro-6-nitroquinoxaline-2,3-dione

CAS Number: 2379-56-8
Molecular Formula: C8H5N3O4
Molecular Weight: 207.15
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 6-Nitro-1,4-dihydroquinoxaline-2,3-dione, 6-Nitroquinoxaline-2,3-dione

Formula: C₈H₅N₃O₄
MW: 207.15 g/mol
MDL Number: MFCD00673833
CAS Number: 2379-56-8
https://us.vwr.com/stibo/web/std.lang.all/02/26/28980226.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR