(2S,5S)-2,5-Diphenylpyrrolidine ≥97.0%

D3186-100MG
TCD3186-100MGEA 953.49 USD
TCD3186-100MG
(2S,5S)-2,5-Diphenylpyrrolidine ≥97.0%
(2S,5S)-2,5-Diphenylpyrrolidine

CAS Number: 295328-85-7
Molecular Formula: C16H17N
Molecular Weight: 223.32
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 50
Specific rotation [a]20/D: -112 deg (C=1, CHCl3)

Formula: C₁₆H₁₇N
MW: 223.32 g/mol
Melting Pt: 50 °C
MDL Number: MFCD07772442
CAS Number: 295328-85-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR