6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic Acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(5-Carboxypentylamino)-7-(N,N-dimethylaminosulfonyl)-2,1,3-benzoxadiazole4-(5-Carboxypentylamino)-7-(N,N-dimethylaminosulphonyl)-2,1,3-benzoxadiazole

D3178-100MG
TCD3178-100MGEA 204 USD
TCD3178-100MG
6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic Acid ≥98.0% (by HPLC, titration analysis)
6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic Acid

Molecular Weight: 356.40
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 154
Molecular Formula: C14H20N4O5S


Other Product Names / Synonyms: 4-(5-Carboxypentylamino)-7-(N,N-dimethylaminosulfonyl)-2,1,3-benzoxadiazole

Formula: C₁₄H₂₀N₄O₅S
MW: 356.40 g/mol
Melting Pt: 154 °C
CAS Number: 1820741-40-9
https://us.vwr.com/stibo/web/std.lang.all/01/78/28980178.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR